missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Bis(2-ethylhexyl) phthalate, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 8003025ML
Specifications
| 117-81-7 | |
| 386°C | |
| C24H38O4 | |
| 25 mL | |
| 390.56 | |
| Viscous Liquid |
| 0.985 g/mL (at 20°C) | |
| 207°C | |
| MFCD00009493 | |
| DEHP; Phthalic acid bis(2-ethylhexyl ester); ′Dioctyl′ phthalate | |
| Selectophore | |
| Selectophore |
Chemical Identifiers
| MFCD00009493 | |
| DEHP; Phthalic acid bis(2-ethylhexyl ester); ′Dioctyl′ phthalate |
Safety and Handling
RTECSNumber : TI0350000
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH