missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Bis(2-ethylhexyl) phthalate, MilliporeSigma™ Supelco™

Catalog No. p-7187947
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018845 5 mL
111018062 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018845 Supplier Merck Emd Millipore Supplier No. 800305ML
Only null left

Chemical Identifiers

MDL Number MFCD00009493
Synonym DEHP; Phthalic acid bis(2-ethylhexyl ester); ′Dioctyl′ phthalate

Specifications

Density 0.985 g/mL (at 20°C)
CAS 117-81-7
Boiling Point 386°C
Flash Point 207°C
Linear Formula C24H38O4
Quantity 5 mL
Formula Weight 390.56
Grade Selectophore
Physical Form Viscous Liquid
Product Line Selectophore
RTECSNumber TI0350000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH