Learn More
Selectophore™ 2-Nitrophenyl octyl ether, ≥99.0%, MilliporeSigma™ Supelco™
$162.22 - $1586.13
Chemical Identifiers
| MFCD00014693 | |
| 1-Nitro-2-octyloxybenzene |
Specifications
| 37682-29-4 | |
| 197°C to 198°C (11 mmHg) | |
| O2 NC6H4 O(CH2)7 CH3 | |
| 5 mL | |
| 251.32 | |
| Selectophore |
| 1.04 g/mL (at 25°C) | |
| 113°C - Closed Cup | |
| MFCD00014693 | |
| 1-Nitro-2-octyloxybenzene | |
| ≥99.0% (GC); ≥99.0% | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH