accessibility menu, dialog, popup

UserName

CAS RN 37682-29-4

IUPAC Name: 1-nitro-2-(octyloxy)benzene
Synonyms: 1-(2-nitrophenoxy)octane
Molecular Weight (g/mol): 251.33
Molecular Formula: C14H21NO3
InChi Key: CXVOIIMJZFREMM-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)