Learn More
Selecto™ Preparative and Classic Column Chromatography Active Silica
60Å gels made from identical starting material, making scale-up easier
$1287.97 - $6561.00
| Catalog Number | Mfr. No. | Particle Size | Quantity | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Particle Size | Quantity | Price | Quantity | ||||
05-719-826
|
Selecto™
162824 |
32 to 63 μm | 25 kg |
Each for $6,561.00
|
|
||||
05-719-866
|
Selecto™
132824 |
32 to 63 μm | 2.5 kg |
Each for $1,287.97
|
|
||||
Description
- pH of 7
- Surface area of 500 to 600m2/g
- Meets needs for standardized initial conditions and high reproducibility
- With 32-63μm particle size is ideal for fast, simple purifying of organic samples
Specifications
| 32 to 63 μm | |
| 7 | |
| Silica Gel |