Learn More
Selecto™ Preparative and Classic Column Chromatography Active Silica DFS Item
60Ă… gels made from identical starting material, making scale-up easier
Supplier:  Selecto™ 132824
 View more versions of this product
Catalog No. 05-719-866
Only null left
Explore available promotions
Description
Description
  • pH of 7
  • Surface area of 500 to 600m2/g
  • Meets needs for standardized initial conditions and high reproducibility
  • With 32-63ÎĽm particle size is ideal for fast, simple purifying of organic samples
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
32 to 63 ÎĽm
7
2.5 kg
Silica Gel
Product Suggestions
Product Suggestions