Learn More
Selecto™ Preparative and Classic Column Chromatography Active Silica
60Å gels made from identical starting material, making scale-up easier
Supplier: Selecto™ 162824
Description
- pH of 7
- Surface area of 500 to 600m2/g
- Meets needs for standardized initial conditions and high reproducibility
- With 32-63μm particle size is ideal for fast, simple purifying of organic samples
Specifications
| 32 to 63 μm | |
| 7 | |
| 25 kg | |
| Silica Gel |