missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application

SecureSeal™ Thermal Adhesive Sealing Film for PCR Application consists of 2.0 mil polyolefin film coated on one side with pressure sensitive acrylate adhesive, which does not interfere with cycle reactions.

$1449.75 - $1530.18

Specifications

Color Transparent
Certifications/Compliance RNase, Dnase and DNA-free
Material Polyolefin
For Use With (Application) PCR
Product Type Sealing Film
View More Specs

Products 2
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
23-045-247
View Documents
Simport™ Scientific
T3291
Non-sterile
Case of 1000 Each for $1,449.75
Only null left
 
23-045-248
View Documents
Simport™ Scientific
T3292
Sterile
Case of 1000 Each for $1,530.18
Only null left
 
Description

Description

  • Low-auto fluorescence
  • Thermostable and functional from -70°C to +100°C
  • Certified RNAse, DNAse and DNA-free
  • DMSO resistant
Specifications

Specifications

Transparent
Polyolefin
Sealing Film
RNase, Dnase and DNA-free
PCR
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'