missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application

Catalog No. 23045248
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
23045248 Sterile
23045247 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23045248 Supplier Simport™ Scientific Supplier No. T3292

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SecureSeal™ Thermal Adhesive Sealing Film for PCR Application consists of 2.0 mil polyolefin film coated on one side with pressure sensitive acrylate adhesive, which does not interfere with cycle reactions.

  • Low-auto fluorescence
  • Thermostable and functional from -70°C to +100°C
  • Certified RNAse, DNAse and DNA-free
  • DMSO resistant

Specifications

Color Transparent
Certifications/Compliance RNase, Dnase and DNA-free
Material Polyolefin
Sterility Sterile
For Use With (Application) PCR
Product Type Sealing Film