missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Second Strand cDNA Synthesis Kit

Catalog No. A48570
Encompass
Change view
Click to view available options
No. of Reactions:
200 Reactions
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A48570 50 Reactions
A48571 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A48570 Supplier Invitrogen™ Supplier No. A48570

Invitrogen™ Second Strand cDNA Synthesis Kit is design to produce double-stranded cDNA using first strand reaction without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.

InvitrogenTM Second Strand cDNA Synthesis Kit is design to produce double-stranded cDNA using first strand reaction without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.

Features of the Second Strand cDNA Synthesis Kit include:

  • Efficient synthesis of full-length double-stranded cDNA
  • Fast—procedure completed in 1 hour
  • Convenient—pre-mixed components
  • Complete—includes residual RNA removal reagents

Applications

  • Full-length double-stranded blunt-end cDNA synthesis for cloning
  • Double-stranded cDNA library construction

Specifications

Content And Storage Contents:
• Second Strand Enzyme Mix, 1 mL
• 5X Second Strand Reaction Mix, 250 μL
• 0.5 M EDTA, pH 8.0, 1 mL
• RNase I, 10 U/μL, 500 μL
• Water, nuclease-free, 3 x 1.25 mL

Storage: -15 to -25°C

Format 50 reactions
Product Type cDNA Synthesis Kit
Quantity 50 reaction
For Use With (Application) Cloning
Final Product Type Double Stranded cDNA
No. of Reactions 50 Reactions
Reaction Format Separate Components
What is the correct reaction temperature that I should use for second strand cDNA synthesis using the Second Strand cDNA Synthesis Kit?

The second strand reaction must be incubated at 16 degrees C to prevent spurious synthesis by DNA polymerase I. At higher temperatures, DNA polymerase I tends to strand-displace rather than nick translate.

When using the Second Strand cDNA Synthesis Kit, what steps do I need to take to properly prepare the second strand reaction mixture?

Dilute the first-strand reaction as outlined in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019413_SecondStrandcDNASynthesisKit_UG.pdf) on page 2 under Second Strand cDNA Synthesis step. Failure to properly dilute the first-strand reaction changes the pH of the second strand reaction and influences the activity of the 3'-5' and 5'-3' exonuclease activities of DNA polymerase I.

When using the Second Strand cDNA Synthesis Kit, how can I avoid incomplete RT inactivation?

Always perform the RT inactivation step after first-strand cDNA synthesis.

I am getting low yield of second strand cDNA product using the Second Strand cDNA Synthesis Kit. What is the reason?

The main reasons are incomplete RT inactivation, improper preparation of the second strand reaction mixture, and incorrect reaction temperature.