missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butylbenzene 99.0+%, TCI America™
$50.33 - $50.33
Chemical Identifiers
| CAS | 135-98-8 |
|---|---|
| Molecular Formula | C10H14 |
| Molecular Weight (g/mol) | 134.222 |
| MDL Number | MFCD00009329 |
| InChI Key | ZJMWRROPUADPEA-UHFFFAOYSA-N |
| Synonym | sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene |
| PubChem CID | 8680 |
| ChEBI | CHEBI:35097 |
| IUPAC Name | butan-2-ylbenzene |
| SMILES | CCC(C)C1=CC=CC=C1 |
Chemical Identifiers
| 135-98-8 | |
| 134.222 | |
| ZJMWRROPUADPEA-UHFFFAOYSA-N | |
| 8680 | |
| butan-2-ylbenzene |
| C10H14 | |
| MFCD00009329 | |
| sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene | |
| CHEBI:35097 | |
| CCC(C)C1=CC=CC=C1 |
Specifications
| 135-98-8 | |
| Colorless | |
| C10H14 | |
| 25 mL | |
| sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene | |
| CCC(C)C1=CC=CC=C1 | |
| 134.222 | |
| CHEBI:35097 | |
| ≥99.0% (GC) | |
| sec-Butylbenzene |
| -83°C | |
| 173°C | |
| MFCD00009329 | |
| 2709 | |
| ZJMWRROPUADPEA-UHFFFAOYSA-N | |
| butan-2-ylbenzene | |
| 8680 | |
| 134.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0011&(3)-0022
RTECSNumber : CY9100000
TSCA : Yes