accessibility menu, dialog, popup

UserName

CH3CH3

CAS RN 135-98-8

IUPAC Name: (butan-2-yl)benzene
Synonyms: sec-butylbenzene secondary butylbenzene s-butylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: ZJMWRROPUADPEA-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)