missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylbenzene 99.0+%, TCI America™

Catalog No. B071425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B071425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B071425ML Supplier TCI America Supplier No. B071425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-98-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009329
InChI Key ZJMWRROPUADPEA-UHFFFAOYSA-N
Synonym sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene
PubChem CID 8680
ChEBI CHEBI:35097
IUPAC Name butan-2-ylbenzene
SMILES CCC(C)C1=CC=CC=C1

Specifications

Melting Point -83°C
Color Colorless
Boiling Point 173°C
Quantity 25 mL
UN Number 2709
Formula Weight 134.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butylbenzene
EINECSNumber (3)-0011&(3)-0022
RTECSNumber CY9100000
TSCA Yes