missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylbenzene, 99+%

Catalog No. p-79217
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107862500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC107862500 Supplier Thermo Scientific Chemicals Supplier No. 107862500
Only null left

CAS: 135-98-8 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135-98-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009329
InChI Key ZJMWRROPUADPEA-UHFFFAOYSA-N
Synonym sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene
PubChem CID 8680
ChEBI CHEBI:35097
IUPAC Name butan-2-ylbenzene
SMILES CCC(C)C1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.8600g/mL
Boiling Point 173°C to 174°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.491
Linear Formula C6H5CH(CH3)C2H5
Quantity 250 mL
Beilstein 05, 414
Specific Gravity 0.86
Merck Index 15, 1552
Solubility Information Solubility in water: 0.015g/L water. Other solubilities: miscible with alcohol,ether and benzene
Formula Weight 134.22
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material sec-Butylbenzene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 205-227-0
RTECSNumber CY9100000
TSCA TSCA

RUO – Research Use Only