missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I048125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I048125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I048125G Supplier TCI America Supplier No. I048125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4426-79-3
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00041108
InChI Key TUFJIDJGIQOYFY-UHFFFAOYNA-N
Synonym sec-butyl isothiocyanate, 2-butyl isothiocyanate, butane, 2-isothiocyanato, isothiocyanic acid sec-butyl ester, isothiocyanic acid, sec-butyl ester, butan-2-isothiocyanate, 2-butylisothiocyanate, s-butyl isothiocyanate, 2-isothiocyanato-butane
PubChem CID 78151
IUPAC Name 2-isothiocyanatobutane
SMILES CCC(C)N=C=S

Specifications

Color Yellow
Boiling Point 159°C
Quantity 25 g
UN Number 1992
Formula Weight 115.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Isothiocyanate