accessibility menu, dialog, popup

UserName

CAS RN 4426-79-3

IUPAC Name: 2-isothiocyanatobutane
Synonyms: butane, 2-isothiocyanato-
Molecular Weight (g/mol): 115.19
Molecular Formula: C5H9NS
InChi Key: TUFJIDJGIQOYFY-UHFFFAOYNA-N
SMILES: CCC(C)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results