missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Alcohol, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B1187200LTBL

 View more versions of this product

Catalog No. 18613086

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Sec-Butyl Alcohol
1
200 L
74.12
Ungraded
78-83-1
94°C to 99.5°C
Drum
0.985

Chemical Identifiers

78-83-1
74.12
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.