missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Alcohol, 98.5%, Spectrum™ Chemical

Catalog No. p-7165167
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613084 100 mL Amber Glass Bottle
18613083 1 L Amber Glass Bottle
18613087 4 L Amber Glass Bottle
18613085 20 L Steel Can
18613086 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613084 Supplier Spectrum Chemical Mfg Cor Supplier No. B1187100ML
Only null left

Chemical Identifiers

CAS 78-83-1
Molecular Weight (g/mol) 74.12

Specifications

CAS Max % 1
Quantity 100 mL
Packaging Amber Glass Bottle
Boiling Point 94°C to 99.5°C
Percent Purity 0.985
Grade Ungraded
Chemical Name or Material Sec-Butyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.