missing translation for 'onlineSavingsMsg'
Learn More

Sebacic Acid 98.0+%, TCI America™

Supplier:  TCI America S0022500G

 View more versions of this product

Catalog No. S0022500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sebacic Acid
134°C
295°C
MFCD00004440
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
C(CCCCC(=O)O)CCCC(=O)O
202.25
CHEBI:41865
≥98.0% (GC,T)
111-20-6
White
C10H18O4
500 g
CXMXRPHRNRROMY-UHFFFAOYSA-N
decanedioic acid
5192
202.25
Crystalline Powder

Chemical Identifiers

111-20-6
202.25
CXMXRPHRNRROMY-UHFFFAOYSA-N
5192
decanedioic acid
C10H18O4
MFCD00004440
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
CHEBI:41865
C(CCCCC(=O)O)CCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0878

RTECSNumber : VS0875000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions