missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sebacic Acid 98.0+%, TCI America™
$34.18 - $75.95
Chemical Identifiers
| CAS | 111-20-6 |
|---|---|
| Molecular Formula | C10H18O4 |
| Molecular Weight (g/mol) | 202.25 |
| MDL Number | MFCD00004440 |
| InChI Key | CXMXRPHRNRROMY-UHFFFAOYSA-N |
| Synonym | sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure |
| PubChem CID | 5192 |
| ChEBI | CHEBI:41865 |
| IUPAC Name | decanedioic acid |
| SMILES | C(CCCCC(=O)O)CCCC(=O)O |
Chemical Identifiers
| C10H18O4 | |
| MFCD00004440 | |
| sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure | |
| CHEBI:41865 | |
| C(CCCCC(=O)O)CCCC(=O)O |
Specifications
| 111-20-6 | |
| White | |
| C10H18O4 | |
| sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure | |
| C(CCCCC(=O)O)CCCC(=O)O | |
| 202.25 | |
| CHEBI:41865 | |
| ≥98.0% (GC,T) | |
| Sebacic Acid |
| 134°C | |
| 295°C | |
| MFCD00004440 | |
| CXMXRPHRNRROMY-UHFFFAOYSA-N | |
| decanedioic acid | |
| 5192 | |
| 202.25 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0878
RTECSNumber : VS0875000
TSCA : Yes