accessibility menu, dialog, popup

UserName

CAS RN 111-20-6

IUPAC Name: decanedioic acid
Synonyms: sebacic acid
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: CXMXRPHRNRROMY-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCC(O)=O
Boiling Point 
  • (4)
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (8)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)