missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek
Ideal for peak identification work
Supplier: Chromatographic RK34483
Description
- CGA 165 connection and can deliver large volumes of sample
- Scotty™ (Air Liquide) 48
- Contents: 48L
- Pressure: 300 psig (21 bar)
- Outlet Fitting: CGA 165
- Weight: 1.75 lb. (0.8kg)
- Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
- DOT Specifications: 39 NRC
All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements
Specifications
| DOT 39 NRC | |
| 10.2 cm | |
| Methane 1% in nitrogen | |
| Ideal for peak identification work | |
| 41 cm | |
| 1.75 lb. | |
| 0.8 kg |
| Methane in Nitrogen | |
| 4 in. | |
| 48 L | |
| 16.25 in. | |
| 300 psi | |
| 12.68 gal. | |
| Air Standard |