missing translation for 'onlineSavingsMsg'
Learn More

Scandium Standard For ICP/MS, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLSC21AM

Catalog No. CLSC21AM


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Scandium Standard
12060-08-1
O3Sc2
ICP-MS
Standard Solution
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
2% HNO3
137.91
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011223
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ICP/ICP-MS Standard
[O--].[O--].[O--].[Sc+3].[Sc+3]
discandium(3+) trioxidandiide
30 mL

Chemical Identifiers

12060-08-1
137.91
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
[O--].[O--].[O--].[Sc+3].[Sc+3]
O3Sc2
MFCD00011223
discandium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Toxic in contact with skin. Causes severe skin burns and eye damage

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only