missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SC 26196

Catalog No. 418910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
418910 10 mg
418950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 418910 Supplier Tocris Bioscience™ Supplier No. 4189/10
Only null left

Selective fatty acid desaturase 2 inhibitor

Selective fatty acid desaturase 2 (FADS2, also known as Δ6 desaturase) inhibitor (IC50 = 0.2 μM in vitro). Displays selectivity over FADS1 and stearoly-CoA 9-desaturase (Δ5 desaturase and SCD-1/Δ9 desaturase, respectively) (IC50 values are >200 μM in vitro). Exhibits anti-inflammatory properties in a mouse edema model. Eliminates CSCs from ovarian cancer cell lines and inhibits sphere formation in vitro.

Chemical Identifiers

CAS 218136-59-5
Molecular Formula C27H29N5

Specifications

Quantity 10 mg
Formula Weight 423.55
Purity >98%
Molecular Weight (g/mol) 423.564
Chemical Name or Material SC 26196
Recommended Storage Store at -20°C