missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ sBADA
Description
sBADA is a green sulfonated BODIPY-FL 3-amino-D-alanine (sBADA) or fluorescent D-amino acid (FFDA). Labels peptidoglycans in live bacteria. Sulfonated form of BADA with increased hydrophilicity and thermostability. Excitation/emission λ ∽490/510 nm.
Specifications
Specifications
| Chemical Name or Material | (R)-2-Amino-3-(3-(5,5-difluoro-7,9-dimethyl-2-sulfo-5H-4λ4,5λ4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanamido)propanoic acid |
| CAS | 2376838-11-6 |
| Quantity | 1 mg |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C17H21BF2N4O6S |
| Purity | 0.95 |