missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SB 225002

A potent, selective and competitive antagonist of the G-protein coupled receptor CXCR2

$499.66

Chemical Identifiers

CAS 182498-32-4
Molecular Formula C13H10BrN3O4
Synonym 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65387MA
View Documents Promotion Details
Thermo Scientific Chemicals
J65387MA
10 mg
Each for $499.66
Only null left
 
Description

Description

SB 225002 is a selective non-peptide CXCR2 inhibitor which suppresses IL-8 induced neutrophil chemotaxis and margination. In addition, SB 225002 reduces MPO activity, IL-1β, MIP-2, and keratinocyte-derived chemokine (KC) levels. Further studies suggest that SB 225002 decreases the expression of vascular endothelial growth factor, inducible NOS (NO synthase), and Cox-2 (cyclooxygenase-2 proteins) in colon tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

182498-32-4
C13H10BrN3O4
1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea

Specifications

Yellow
A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2
C13H10BrN3O4
1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea
Soluble to 100mM in DMSO and to 50mM in ethanol
Solid
OC1=C(NC(=O)NC2=CC=CC=C2Br)C=CC(=C1)[N+]([O-])=O
352.14
-30°C to -10°C
182498-32-4
352.10g/mol
Moisture Sensitive
Signal Transduction Reagent
MQBZVUNNWUIPMK-UHFFFAOYSA-N
3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only