CAS RN 182498-32-4
IUPAC Name:
3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Synonyms:
3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Molecular Weight (g/mol):
352.14
Molecular Formula:
C13H10BrN3O4
InChi Key:
MQBZVUNNWUIPMK-UHFFFAOYSA-N
SMILES:
OC1=C(NC(=O)NC2=CC=CC=C2Br)C=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals SB 225002
CAS: 182498-32-4 Molecular Formula: C13H10BrN3O4 Synonym: 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea