missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 6 Centrifugal Concentrators

Catalog No. p-4187761
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
0.2 μm
Quantity:
100/Pk.
25/Pk.
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14558463 10 kDa 25/Pk.
14558464 10 kDa 100/Pk.
14558465 5 kDa 25/Pk.
14558466 5 kDa 100/Pk.
14558467 30 kDa 25/Pk.
14558468 30 kDa 100/Pk.
14558469 50 kDa 25/Pk.
14558470 50 kDa 100/Pk.
14558472 100 kDa 100/Pk.
14558471 100 kDa 25/Pk.
14558473 300 kDa 25/Pk.
14558474 300 kDa 100/Pk.
14558478 0.2 μm 100/Pk.
14558480 3 kDa 100/Pk.
14558479 3 kDa 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 14558463 Supplier Sartorius Supplier No. VS0601
Only null left

Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.

Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 10 kDa
Sample Volume 6 mL
Sterility Non-sterile
Filtration Area 2.58 cm2
Housing Material Polycarbonate
Quantity 25/Pk.
Volume (Metric) 6 mL
Length (Metric) 122 mm
Show More Show Less