Learn More
Sartorius Vivaspin™ 500 Centrifugal Concentrators
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
Supplier: Sartorius VS0172
Description
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
- Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
- Concentrate recoveries of > 90% can be reached in less than 30 minutes
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
- High-flux PES membranes for ultra-fast concentration
- Convenient concentrate recovery by pipetting
- Clear housing with volume graduation for easy reading/monitoring
Chemical Identifiers
| Single-use | |
| Centrifuge | |
| 500 mL | |
| 64.5m m2 | |
| 50 mm |
| Ultrafiltration | |
| PES | |
| Non-sterile | |
| Polycarbonate |
Specifications
| Single-use | |
| Centrifuge | |
| 0.2 μm | |
| 500 mL | |
| 64.5m m2 | |
| 100/Pk. | |
| 50 mm |
| Ultrafiltration | |
| PES | |
| 0.2 μm | |
| Non-sterile | |
| Polycarbonate | |
| 500 μL |