Learn More
Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane
Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.
$215.47 - $753.68
Specifications
| Disposable | Single-use |
|---|---|
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Centrifuge |
| Material (Membrane) | PES |
| Sterility | Non-sterile |
| Catalog Number | Mfr. No. | MWCO | Quantity | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | MWCO | Quantity | Price | Quantity | ||||
|
14-558-419
|
Sartorius
VS0211 |
5 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-418
|
Sartorius
VS0202 |
10 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
|
14-558-421
|
Sartorius
VS0221 |
30 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-417
|
Sartorius
VS0201 |
10 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-420
|
Sartorius
VS0212 |
5 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
|
14-558-422
|
Sartorius
VS0222 |
30 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
|
14-558-423
|
Sartorius
VS0231 |
50 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-424
|
Sartorius
VS0232 |
50 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
|
14-558-425
|
Sartorius
VS0241 |
100 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-426
|
Sartorius
VS0242 |
100 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
|
14-558-428
|
Sartorius
VS0252 |
300 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
|
14-558-429
|
Sartorius
VS0261 |
1000 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-433
|
Sartorius
VS0291 |
3 kDa | 25/Pk. |
Pack of 25 Each for $215.47
|
|
||||
|
14-558-432
|
Sartorius
VS0272 |
100/Pk. |
Pack of 100 Each for $753.68
|
|
|||||
|
14-558-434
|
Sartorius
VS0292 |
3 kDa | 100/Pk. |
Pack of 100 Each for $753.68
|
|
||||
Description
Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.
Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.
Twin vertical membranes and optimal design for fast, safe concentration
- Patented twin vertical membrane design for ultra-fast processing speeds
- In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5-30 minutes
- Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
- Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
- High-flux PES membranes feature very low protein binding and low extractables
Designed for maximum flexibility and sample recovery
- Low internal surface and membrane area for superior recoveries from very dilute solutions
- Two methods for easy concentrate recovery:
- Directly pipet the concentrate from the dead stop pocket in the concentrator
- Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
- Both methods provide near-total concentrate recovery of greater than 90%
- Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
- Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes
Specifications
| Single-use | |
| Centrifuge | |
| Non-sterile | |
| Concentration, desalting and buffer applications | |
| Centrifugal Concentrator | |
| 126 mm |
| Ultrafiltration | |
| PES | |
| 1.3 cm2 | |
| Polycarbonate | |
| 2 mL | |
| 17.8 mm |