missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivaspin™ 2 Centrifugal Concentrator with Hydrosart™ Regenerated Cellulose (RC) Membrane

Catalog No. 14558442
Change view
Click to view available options
MWCO:
10 kDa
2 kDa
30 kDa
5 kDa
Quantity:
100 Pk.
25 Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14558442 2 kDa 100 Pk.
14558435 10 kDa 25 Pk.
14558436 10 kDa 100 Pk.
14558437 5 kDa 25 Pk.
14558438 5 kDa 100 Pk.
14558441 2 kDa 25 Pk.
14558440 30 kDa 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14558442 Supplier Sartorius™ Supplier No. VS02H92
Only null left

Vivaspin™ 2 Centrifugal Concentrators are disposable ultrafiltration units for concentration of macromolecules. With a broad range of molecular weight cut-offs (MWCO) and membrane materials they are suitable for various substrates, including proteins, viruses, nucleic acids, and nanoparticles.

Vivaspin concentrators are designed for the concentration or diafiltration of initial sample volumes up to 3 mL and can be used with swinging bucket or fixed angle rotors that accept 15 mL centrifuge tubes. Vivaspin's patented vertical membrane design and thin channel filtration chamber (US 5,647,990) minimize membrane fouling and provide high-speed concentration for even particle-laden solutions. The speed of Vivaspin products combined with low internal surface and membrane area allow superior recovery from very dilute solutions. The Vivaspin 2 also offers flexibility for process optimization: one can directly pipette the concentrate from the dead stop pocket (built into the bottom of the concentrator) or reverse-spin the concentrate into the recovery cap (which can be sealed for storage). Either method results in near-total concentrate recovery.

  • High retentate recovery >90%
  • Vertical membrane design for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Optional re-spin for concentrate recovery
  • Volume graduation for easy reading/monitoring

Specifications

Disposable Single-use
Filter Type Membrane Filter
For Use With (Equipment) Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
Material (Membrane) Hydrosart
MWCO 2 kDa
Sample Volume Up to 3 mL
Sterility Non-sterile
Quantity 100 Pk.
Type Centrifugal Concentrator
Volume (Metric) 3 mL
Length (English) 5 in.
Length (Metric) 126 mm
Show More Show Less