missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivaspin™ 2 Centrifugal Concentrator with Hydrosart™ Regenerated Cellulose (RC) Membrane

Vivaspin™ 2 Centrifugal Concentrators are disposable ultrafiltration units for concentration of macromolecules. With a broad range of molecular weight cut-offs (MWCO) and membrane materials they are suitable for various substrates, including proteins, viruses, nucleic acids, and nanoparticles.

Supplier:  Sartorius™ VS02H22

 View more versions of this product

Catalog No. 14558440


Only null left

Description

Description

Vivaspin concentrators are designed for the concentration or diafiltration of initial sample volumes up to 3 mL and can be used with swinging bucket or fixed angle rotors that accept 15 mL centrifuge tubes. Vivaspin's patented vertical membrane design and thin channel filtration chamber (US 5,647,990) minimize membrane fouling and provide high-speed concentration for even particle-laden solutions. The speed of Vivaspin products combined with low internal surface and membrane area allow superior recovery from very dilute solutions. The Vivaspin 2 also offers flexibility for process optimization: one can directly pipette the concentrate from the dead stop pocket (built into the bottom of the concentrator) or reverse-spin the concentrate into the recovery cap (which can be sealed for storage). Either method results in near-total concentrate recovery.

  • High retentate recovery >90%
  • Vertical membrane design for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Optional re-spin for concentrate recovery
  • Volume graduation for easy reading/monitoring
Specifications

Chemical Identifiers

Single-use
Hydrosart
Non-sterile
3 mL
126 mm
Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
Up to 3 mL
Centrifugal Concentrator
5 in.

Specifications

Single-use
Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
30 kDa
Non-sterile
Centrifugal Concentrator
5 in.
Ultrafiltration
Hydrosart
Up to 3 mL
100 Pk.
3 mL
126 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions