missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 15R Centrifugal Concentrators

Catalog No. 14558499
Change view
Click to view available options
MWCO:
10 kDa
2 kDa
30 kDa
5 kDa
Quantity:
12/Pk.
48/Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14558499 2 kDa 48/Pk.
14558492 10 kDa 12/Pk.
14558493 10 kDa 48/Pk.
14558496 30 kDa 12/Pk.
14558494 5 kDa 12/Pk.
14558497 30 kDa 48/Pk.
14558495 5 kDa 48/Pk.
14558498 2 kDa 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14558499 Supplier Sartorius Supplier No. VS15RH92

Features a Hydrosart™ modified regenerated cellulose membrane. Sartorius Vivaspin 15R Centrifugal Concentrators are ideal when extremely high recovery rate and very low adsorption is required, e.g. applications such as concentration and desalting of immunoglobulin fractions. Available in a range of MWCO.

The latest member of the Vivaspin product family
  • With all the unique features of Sartorius Stedim Biotech centrifugal concentrators
  • Conical bottom, 30 x 116mm tubes that target a volume of 2 to 15mL
  • Patented vertical twin membrane for extremely short concentration time (30-fold in 15 min.)
  • Safe sample concentration due to dead-stop volume
  • Features the Hydrosart™ regenerated cellulose membrane
  • Highest recovery at low absorption (95-98%)
  • Very small holdup volume (< 20μl)
  • Crystal clear housing with volume graduations
Rotor Compatibility
  • Designed to fit into rotors that can accommodate Falcon 50mL conical bottom tubes
    • Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors
    • Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
    • Multifuge 3 S-R with 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 ml conical bottom tubes
  • These devices are not designed to fit into rotors that only accept round bottom 29 mm x 105 mm tubes (e.g. Sorvall SS34 Beckmann JA 20)
    • If your rotor accepts only 29 mm x 105 mm round bottom tubes, please use the Vivaspin 15, which can be used in either round bottom or conical centrifuge tubes

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) Hydrosart
MWCO 2 kDa
Sample Volume 15 mL
Sterility Non-sterile
Filtration Area 3.87 cm2
Housing Material Polycarbonate
Quantity 48/Pk.
Type Centrifugal Concentrator
Volume (Metric) 12.5 mL
Length (Metric) 116 mm
Show More Show Less