missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivapure™ S Mini H IEX Spin Columns
Description
Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel.
- Fast centrifugal spin columns
- Easy and ready-to-use
- Ideal for parallel processing
- Sample preparation prior to 1-D or 2-D PAGE
- Purification scouting of an unknown protein
- Removal of endotoxins from monoclonal antibodies
- Preparation of heme moiety from heme containing protein for functional analysis
- Polishing of His-tagged proteins
- General protein purification
- Detergent removal from protein solutions
- Purification of antibodies from serum, ascites or tissue culture supernatant
- Establishing a purification/ pre-purification protocol for a given protein
- HPLC/FPLC sample preparation
- Purification of membrane-bound proteins
Specifications
Specifications
| Bed Volume | 240 μL |
| Material | Polypropylene |
| Pore Size | 3 to 5 μm |
| Support Type | Stabilized regenerated Cellulose |
| Particle Size | Not Applicable |
| Form | Agarose Bead |
| Quantity | 24/Pk. |
| For Use With (Application) | Sample fractionation, purification condition scouting, small scale purification |
| Type | Spin Column |
| Sample Volume | 24 x 0.5 mL |
| Show More |