missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivapure™ LentiSELECT 500 Purification and Concentration Kit

Catalog No. 14558565
Change view
Click to view available options
Sample Size:
500 mL of Lentivirus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size
14558565 500 mL of Lentivirus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558565 Supplier Sartorius Supplier No. VS LVPQ500

Fast purification of up to 3 x109 viral particles

Vivapure™ LentiSELECT 500 is optimally suited for VSV-G pseudotyped lentivirus purification from up to 500mL cell culture and contains all reagents and devices necessary for purifying up to 2-5 x 109 infective particles.

The whole purification procedure is simply operated by a laboratory pump, which min imizes hands-on time. Unlike conventional purification methods as ultracentrifugation, Vivapure LentiSELECT 500 offers a fast and simple solution for purifying VSV-G pseudotyped lentiviruses making expensive purification equipment like ultracentrifuges redundant.

Typically, for a normal yielding vector, a 500mL cell culture purified using this method will yield up to 2-5 x 109 infective particles in 1mL (total volume 1mL).

  • Fast
  • Non-toxic
  • Better purity than pelleting by ultracentrifugation
  • Convienient
  • Cost effective

Specifications

Sample Size 500 mL of Lentivirus
Purification Target 40 to 1000 mL Cell Cultures
For Use With (Application) VSV-G Pseudotyped Lentiviral Vectors