missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivapure™ D Mini H IEX Spin Column

Catalog No. 14558551
Change view
Click to view available options
Capacity (Metric):
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14558551 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558551 Supplier Sartorius™ Supplier No. VSIX01DH24

Ready-to-use centrifugal devices based on Sartobind Membrane Adsorber technology as their chromatography matrix

Vivapure Mini Ion Exchange (IEX) spin columns are centrifugal devices, incorporating Sartobind Membrane Adsorber technology as their chromatography matrix. Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel. VIvapure Mini is available in Diethylamine (D) membrane chemistry.
Typical Applications

  • Sample preparation prior to 1-D or 2-D PAGE
  • Purification scouting of an unknown protein
  • Removal of endotoxins from monoclonal antibodies
  • Preparation of heme moiety from heme containing protein for functional analysis
  • Polishing of His-tagged proteins
  • General protein purification
  • Detergent removal from protein solutions
  • Purification of antibodies from serum, ascites or tissue culture supernatant
  • Establishing a purification/ pre-purification protocol for a given protein
  • HPLC/FPLC sample preparation
  • Purification of membrane-bound proteins
Estimated protein binding capacity: 4mg

  • Fast centrifugal spin columns
  • Easy and ready-to-use
  • Binding capacity 4mg protein
  • Ideal for parallel processing

Specifications

Bed Volume 240 μL
Capacity (Metric) 0.4 mL
Binding Property High
Material Polypropylene
Pore Size 3 to 5 μm
Support Type Stabilized regenerated Cellulose
Quantity 24/Pk.
For Use With (Application) Paralell processing
Type Spin Column