missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivapure™ AdenoPACK 500 Kit

Catalog No. 14558550
Change view
Click to view available options
Endotoxin Concentration:
<0.025 EU/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Endotoxin Concentration
14558550 <0.025 EU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558550 Supplier Sartorius Supplier No. VS AVPQ501

Pump driven Kit for larger volumes

Vivapure™ AdenoPACK 500 is the direct upscale kit to the AdenoPACK 100, for adenovirus purification. In only 2 hours up to 3 x 1013 adenovirus particles are purified and concentrated from 500 ml cell culture. This completely ready-to-use kit is conveniently operated by a laboratory pump, offering optimal flow control and minimal hands-on time. This easy to use product replaces lengthy and inefficient cesium chloride density gradient methods.

  • Pump driven kit
  • Easy and controlled purification of Adenovirus from large cell culture volumes
  • Virus purification based on membrane adsorber technology
  • High-speed process completed in about 2 hours
  • Convenient, easy to use alternative to CsCl density gradients
  • Able to process 10 to 25 plates (up to 500 ml)
  • Flexible, most cost-effective kit in the market for purifying 1-3 x 1013 viral particles

Specifications

Endotoxin Concentration <0.025 EU/mL
Sample Size 500 mL of Adenovirus supernatants
Purification Target Adenovirus
Content And Storage -20°C
For Use With (Application) AdenoPACK 100