missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ Models Accessories: Flow Restrictors

Catalog No. 14558337
Change view
Click to view available options
For Use With (Equipment):
Vivaflow 50 and 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14558337 Vivaflow 50 and 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558337 Supplier Sartorius Supplier No. VFA035

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flow Restrictors for Vivaflow™ 50 and 200.

  • Manage and maintain flow

Specifications

Color Pink
Filter Type Ultrafiltration
Material PVC
Pore Size 0.6 mm
Sterility Non-sterile
Quantity 6 Pk.
Type Flow Restrictor
For Use With (Equipment) Vivaflow 50 and 200