missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaflow™ 50R Crossflow Casettes
Unique, Plug and Play laboratory crossflow devices for concentrating samples like viruses and antibodies
Supplier: Sartorius VF05H2
Includes: Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
Description
- Reusable device with low-binding Hydrosart™ (modified regenerated cellulose) membrane
- For concentrating up to 1L samples
- Active membrane area per device is 50cm2
- One unit comes with all necessary accessories for running device with laboratory pump and size 16 pump head
- For speeding up concentration, two cassettes can be run simultaniously
- Acrylic main housing and flow channel
- Polypropylene membrane support
- Nylon fittings and PVC tubing
- Compatible with 1M NaOH for cleaning and depyrogenation
- High flow rate and recovery
- L x W x H: 100x 24 x 100mm
- Pump flow rate: 200 to 400mL/min
- Maximum pressure: 45psi
- Maximum temperature: 60°C
- Antibody/recombinant protein concentration/diafiltration in biopharma research
- Concentration of small production lots of proteins for diagnostic purposes
- Concentration of viruses from cell culture supernatants or of environmental samples
- Nanoparticle concentration
Specifications
| Ultrafiltration | |
| Labscale TFF System | |
| Cellulose | |
| 1 L | |
| Concentrating viruses and antibodies | |
| Protein, Virus, Nanoparticles | |
| 1 L |
| Cassette | |
| Acrylic | |
| 30 kDa | |
| Non-sterile | |
| Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings | |
| Crossflow Cassette |