missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ 50R Crossflow Casettes

Unique, Plug and Play laboratory crossflow devices for concentrating samples like viruses and antibodies

Supplier:  Sartorius VF05H0

 View more versions of this product

Catalog No. 14-558-021


Only null left
Explore available promotions

Includes: Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings

Description

Description

  • Reusable device with low-binding Hydrosart™ (modified regenerated cellulose) membrane
  • For concentrating up to 1L samples
  • Active membrane area per device is 50cm2
  • One unit comes with all necessary accessories for running device with laboratory pump and size 16 pump head
  • For speeding up concentration, two cassettes can be run simultaniously
  • Acrylic main housing and flow channel
  • Polypropylene membrane support
  • Nylon fittings and PVC tubing
  • Compatible with 1M NaOH for cleaning and depyrogenation
  • High flow rate and recovery
  • L x W x H: 100x 24 x 100mm
  • Pump flow rate: 200 to 400mL/min
  • Maximum pressure: 45psi
  • Maximum temperature: 60°C
  • Antibody/recombinant protein concentration/diafiltration in biopharma research
  • Concentration of small production lots of proteins for diagnostic purposes
  • Concentration of viruses from cell culture supernatants or of environmental samples
  • Nanoparticle concentration
Specifications

Specifications

Ultrafiltration
Labscale TFF System
Cellulose
1 L
Concentrating viruses and antibodies
Protein, Virus, Nanoparticles
1 L
Cassette
Acrylic
10 kDa
Non-sterile
Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
Crossflow Cassette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions