missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ 200 Module

Catalog No. 14556110
Change view
Click to view available options
Volume (Metric) Hold Up:
5.3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Hold Up
14556110 5.3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14556110 Supplier Sartorius Supplier No. VF20H9

Vivaflow™ 200 Module is a plug and play laboratory cross flow cassette. Re-usable. Optimal for cell culture supernatant and virus concentration. High flow rate and recovery.

  • Vivaflow™ 200 is a ready to use and reusable laboratory crossflow cassette for concentrating up to 5 L samples.
  • The membrane surface per device is 200 cm2.
  • One unit comes with all necessary accessories for running the device with a laboratory pump and a size 16 pump head.
  • For speeding up the concentration, two cassettes can be run simultaneously.
  • Vivaflow™ 200 is optimally suited for applications like cell culture supernatant concentration, virus concentration and water concentration e.g. in fish breeding facilities.
  • This economical product offers a standard of ease of use, reliability and flexibility unrivalled by any laboratory filtration system in the market.
  • The thin-channel, flip-flow recirculation path of the modules provides high crossflow velocities with minimum pump speed requirements.
  • A single 200 cm2 module typically concentrates 1 L by a factor of 50 in less than 30 min. Alternatively, two Vivaflow™ 200 Modules can be used in parallel for concentration of 5 L in under 75 minutes.
  • Near total recovery of the concentrate is achieved with a single final rinse.
  • Vivaflow™ 200 can be easily cleaned and re-used several times.
  • All accessories for sample concentration, except a peristaltic pump with a pump head for size 16 tubing, are included in the package for using a single cassette.
  • A 500 mL sealed diafiltration reservoir can be added to the system for rapid and convenient buffer exchange.

Equipment required:

  • The standard Vivaflow™ 200 package comes with tubing, pressure indicator, flow restrictor and high pressure pump tubing.
  • All you need is a peristaltic pump with a pump head capable of handling 6.4 mm OD tubing (size 16).

Running 2 Vivaflow™ 200 cassettes in parallel:

  • For speeding up the concentration, two Vivaflow™ 200 cassettes can easily be connected using a Y connector which is preassembeled with the correct tubing sizes and fittings (size 16 tubing for connecting the two Vivaflow™ 200 units, size 15 tubing for the pump).
  • Please note that you need a size 15 pump head for running two Vivaflows™ in parallel.
  • High flow rate and recovery.
  • Clear housing for process monitoring.
  • Ready to use single cassettes.
  • Easy set up of 2 cassettes for use in parallel.
  • Re-usable product.

Specifications

Certifications/Compliance ISO 9001
Disposable Reusable
Filter Type TFF/Crossflow
Material Acrylic
Material (Membrane) Hydrosart
Sterility Non-sterile
Filtration Area 200 cm2
For Use With (Application) Concentration desalting and buffer exchange
Max. Pressure 4 bar (58 psi)
Quantity 1 Ea.
Temperature (English) Max. 60°C (140°F)
Type TFF Cassette
Volume (Metric) 0.5 to 5 L
Volume (Metric) Hold Up 5.3 mL
Height (English) 5.43 in.
Height (Metric) 13.8 cm
Sample Volume 2.5 L
Show More Show Less