missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 2 DNA Concentrator

Catalog No. 14558380
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
2 kDa
30 kDa
50 kDa
Quantity:
100 Pack
25 Pack
500 Pack
Product Line:
Hydrosart
Vivacon 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity Product Line
14558380 2 kDa 100 Pack Vivacon 2
14558361 10 kDa 100 Pack Hydrosart
14558364 30 kDa 25 Pack Hydrosart
14558365 30 kDa 100 Pack Hydrosart
14558370 50 kDa 100 Pack Vivacon 2
14558374 100 kDa 100 Pack Vivacon 2
14558376 100 kDa 500 Pack Vivacon 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14558380 Supplier Sartorius Supplier No. VN02H92

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • Graduation and MWCO printed on device
  • Complete and highly reproducible concentrate recovery

Specifications

Concentration 50ng/mL
Concentration Time 120 min.
Filter Type Ultrafiltration
Material Polycarbonate
Material (Membrane) Cellulose
MWCO 2 kDa
Sterility Non-sterile
Filtration Area 0.95 cm2
For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
Max. Force 7500xG
Product Line Vivacon 2
Quantity 100 Pack
Sample Type DNA, peptide
Type DNA Concentrator
Volume (Metric) 2 mL
Volume (Metric) Hold Up 10 μL
Dimensions (L x W) 4.92 x 0.62 in. (12.5 x 1.6cm)
Length (English) 4.92 in.
Length (English) Back Spin 4.52 in.
Length (Metric) 12.5 cm
Length (Metric) Back Spin 11.5 cm
Width (English) 0.62 in.
Width (Metric) 1.6 cm
For Use With (Equipment) Centrifuge
Sample Volume 2 mL
Show More Show Less