Learn More
Sartorius Glass Filtration Holder
Catalog No. 14555547
Change view
Click to view available options
Type:
Millex Samplicity Filter
Vacuum Filter Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14555547 Millex Samplicity Filter
14555544 Vacuum Filter Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14555547 Supplier Sartorius Supplier No. 16315
This glass vacuum filtration holder has been designed for particle testing, clarification and sterile filtration with the use of 24 or 25 mm filters.
  • Vaccuum Filter Holder is delivered with a glass frit filter support that ensures a uniform distribution of the retained particles; therefore it is recommended when the residue on the filter surface is of interest.
  • Millex Filter is delivered with a PTFE-coated screen filter support that has the benefit to be easy to clean. Therefore it is recommended when the filtrate is required or when liquids that are difficult to remove from glass supports must be examined.
Specifications
Disposable Reusable
Material Glass
Sterility Sterile
Filtration Area 3 cm2
Quantity Each
Type Millex Samplicity Filter
Volume (Metric) 30 mL
Diameter (Metric) Membrane 2.5 cm
For Use With (Equipment) Vacuum Filter Holder, Manifold System