missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartoclear Dynamics™ Lab V Filtration Kit
Description
Comprised of:
- A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
- A bottle, with graduation and writing field
- A tube connector for vacuum connection (for stand-alone filtration)
- A screw cap for storage of the filtrate (individually wrapped to maintain sterility) history
Benefits:
- Kits for a single-step mammalian cell culture harvesting
- Ready to use
- Fast and effortless
Specifications
Specifications
| Filter Type | Diatomaceous Earth, 5 g |
| Product Type | Vacuum Filtration Kit |
| Quantity | 24/Cs. |
| Includes | 2 x 180E04---------E (24 x Sartolab™ RF 500; 0.22 μm; PES; single-packaged; sterile); 1 x SDLKG-05.0------2 (24 x pouches of filter aid, 5 g) |
| Capacity | 500 mL |