Learn More
Sarcosine ethyl ester hydrochloride, 98+%
CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol
Supplier: Thermo Scientific Chemicals A1311236
Description
Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.
Solubility
Soluble in water 0.1 g/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| Sarcosine ethyl ester hydrochloride | |
| 122°C to 126°C | |
| CH3NHCH2CO2CH2CH3·HCl | |
| 500 g | |
| Hygroscopic | |
| Soluble in water 0.1g/mL. | |
| [H+].[Cl-].CCOC(=O)CNC | |
| 153.61 | |
| 153.61 |
| 52605-49-9 | |
| C5H12ClNO2 | |
| MFCD00012506 | |
| 3911182 | |
| sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride | |
| NIDZUMSLERGAON-UHFFFAOYSA-N | |
| ethyl 2-(methylamino)acetate;hydrochloride | |
| 171173 | |
| ≥98% |
Chemical Identifiers
| 52605-49-9 | |
| 153.61 | |
| NIDZUMSLERGAON-UHFFFAOYSA-N | |
| 171173 |
| C5H12ClNO2 | |
| MFCD00012506 | |
| sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride | |
| [H+].[Cl-].CCOC(=O)CNC |
Safety and Handling
EINECSNumber : 258-037-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only