accessibility menu, dialog, popup

UserName

HClH3COONHCH3

CAS RN 52605-49-9

IUPAC Name: hydrogen ethyl 2-(methylamino)acetate chloride
Synonyms: hydrogen ethyl 2-(methylamino)acetate chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: NIDZUMSLERGAON-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=O)CNC
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)