missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine ethyl ester hydrochloride, 98+%

Catalog No. p-7020969
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1311214 25 g
AAA1311222 100 g
AAA1311236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1311214 Supplier Thermo Scientific Chemicals Supplier No. A1311214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol

Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.

Solubility
Soluble in water 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 52605-49-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012506
InChI Key NIDZUMSLERGAON-UHFFFAOYSA-N
Synonym sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
PubChem CID 171173
SMILES [H+].[Cl-].CCOC(=O)CNC

Specifications

Melting Point 122°C to 126°C
Linear Formula CH3NHCH2CO2CH2CH3·HCl
Quantity 25 g
Beilstein 3911182
Sensitivity Hygroscopic
Solubility Information Soluble in water 0.1g/mL.
IUPAC Name hydrogen ethyl 2-(methylamino)acetate chloride
Formula Weight 153.61
Percent Purity ≥98%
Chemical Name or Material Sarcosine ethyl ester hydrochloride
EINECSNumber 258-037-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only