missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sarcosine, 98%
CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol
$103.48 - $1,194.18
Chemical Identifiers
| CAS | 107-97-1 |
|---|---|
| Molecular Formula | C3H7NO2 |
| Molecular Weight (g/mol) | 89.09 |
| MDL Number | MFCD00004279 |
| InChI Key | FSYKKLYZXJSNPZ-UHFFFAOYSA-N |
| Synonym | N-Methylglycine |
| PubChem CID | 1088 |
| ChEBI | CHEBI:15611 |
| IUPAC Name | 2-(methylamino)acetic acid |
| SMILES | CNCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC157741000
|
Thermo Scientific Chemicals
157741000 |
100 g | Plastic bottle |
Each for $103.48
|
|
||||
|
AC157745000
|
Thermo Scientific Chemicals
157745000 |
500 g | Plastic bottle |
Each for $304.32
|
|
||||
|
AC157740025
|
Thermo Scientific Chemicals
157740025 |
2.5 kg | Plastic bottle |
Each for $1,194.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C3H7NO2 | |
| MFCD00004279 | |
| N-Methylglycine | |
| CHEBI:15611 | |
| CNCC(O)=O |
Specifications
| Colorless or White to Yellow | |
| Crystals or Powder | |
| 98% | |
| 2% max. | |
| MFCD00004279 | |
| FSYKKLYZXJSNPZ-UHFFFAOYSA-N | |
| 2-(methylamino)acetic acid | |
| 1088 | |
| 89.09 | |
| Sarcosine |
| 107-97-1 | |
| Authentic | |
| C3H7NO2 | |
| CH3NHCH2CO2H | |
| N-Methylglycine | |
| CNCC(O)=O | |
| 89.09 | |
| CHEBI:15611 | |
| ≥97.5% (total base) |
RUO – Research Use Only