missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine, 98%

Catalog No. AC157745000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157745000 500 g Plastic bottle
AC157741000 100 g Plastic bottle
AC157740025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157745000 Supplier Thermo Scientific Chemicals Supplier No. 157745000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym N-Methylglycine
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O

Specifications

Color Colorless or White to Yellow
Physical Form Crystals or Powder
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3NHCH2CO2H
Water 2% max.
Formula Weight 89.09
Percent Purity ≥97.5% (total base)
Chemical Name or Material Sarcosine
Show More Show Less

RUO – Research Use Only