missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine, 98%

CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol

Supplier:  Thermo Scientific Chemicals 157740025

Quantity 2.5 kg
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC157740025


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-97-1
89.09
FSYKKLYZXJSNPZ-UHFFFAOYSA-N
1088
2-(methylamino)acetic acid
C3H7NO2
MFCD00004279
N-Methylglycine
CHEBI:15611
CNCC(O)=O

Specifications

Sarcosine
107-97-1
2.5 kg
Plastic bottle
C3H7NO2
2% max.
N-Methylglycine
CNCC(O)=O
89.09
CHEBI:15611
≥97.5% (total base)
Colorless or White to Yellow
Crystals or Powder
Authentic
98%
CH3NHCH2CO2H
MFCD00004279
FSYKKLYZXJSNPZ-UHFFFAOYSA-N
2-(methylamino)acetic acid
1088
89.09
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions